missing translation for 'onlineSavingsMsg'
Learn More

4-Nitrosopiperazine-1-ethanol, TRC

Product Code. 30481198
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30481198 500 mg 500mg
30466134 50 mg 50mg
30473870 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30481198 Supplier Toronto Research Chemicals Supplier No. TRCN481260500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 48121-20-6
Molecular Formula C6 H13 N3 O2
Molecular Weight (g/mol) 159.186
Synonym 4-Nitroso-1-piperazineethanol, 2-(4-Nitroso-piperazin-1-yl)-ethanol, 2-(4-Nitrosopiperazin-1-yl)ethan-1-ol, 4-Nitrosopiperazine-1-ethanol
IUPAC Name 2-(4-nitrosopiperazin-1-yl)ethanol
SMILES OCCN1CCN(CC1)N=O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C6H13N3O2/c10-6-5-8-1-3-9(7-11)4-2-8/h10H,1-6H2
Formula Weight 159.1008
Chemical Name or Material Aripiprazole Impurity 30
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.