CAS RN 48121-20-6
CAS RN 48121-20-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
4-Nitrosopiperazine-1-ethanol, TRC
CAS: 48121-20-6 Molecular Formula: C6 H13 N3 O2 Molecular Weight (g/mol): 159.186 Synonym: 4-Nitroso-1-piperazineethanol,2-(4-Nitroso-piperazin-1-yl)-ethanol,2-(4-Nitrosopiperazin-1-yl)ethan-1-ol,4-Nitrosopiperazine-1-ethanol IUPAC Name: 2-(4-nitrosopiperazin-1-yl)ethanol SMILES: OCCN1CCN(CC1)N=O
2
4-Nitroso-1-piperazineethanol, Mikromol
Discover Mikromol- your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.