missing translation for 'onlineSavingsMsg'
Learn More

4-Nitro-L-phenylalanine monohydrate, 98%

Product Code. 11383326
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11383326

Brand: Thermo Scientific Alfa Aesar L12709.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 207591-86-4 | C9H12N2O5 | 228.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207591-86-4
Molecular Formula C9H12N2O5
Molecular Weight (g/mol) 228.204
MDL Number MFCD00051221
InChI Key OLZDHJRNUIXKLU-QRPNPIFTSA-N
Synonym 4-nitro-l-phenylalanine monohydrate, 4-nitro-l-phenylalanine hydrate, s-2-amino-3-4-nitrophenyl propanoic acid hydrate, h-phe 4-no2-oh.h2o, 2s-2-amino-3-4-nitrophenyl propanoic acid hydrate, 4-nitro-l-phenylalanin hydrate, phenylalanine, 4-nitro-hydrate, l-phenylalanine, 4-nitro-, monohydrate, 4-nitro-l-phenylalnine, pubchem12061
PubChem CID 2734751
IUPAC Name (2S)-2-amino-3-(4-nitrophenyl)propanoic acid;hydrate
SMILES C1=CC(=CC=C1CC(C(=O)O)N)[N+](=O)[O-].O

Specifications

Melting Point 236°C to 239°C (decomposition)
Beilstein 2809673
Quantity 5 g
Formula Weight 228.21 (210.19 Anhydrous)
Optical Rotation +6.5° (c=1 in 5N HCl)
Percent Purity 98%
Chemical Name or Material 4-Nitro-L-phenylalanine monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.