accessibility menu, dialog, popup

UserName

CAS RN 207591-86-4

OHONH2NOOH2O

CAS RN 207591-86-4

IUPAC Name: 2-amino-3-(4-nitrophenyl)propanoic acid hydrate
Synonyms: p-nitrophenylalanine hydrate
Molecular Weight (g/mol): 228.2
Molecular Formula: C9H12N2O5
InChi Key: OLZDHJRNUIXKLU-UHFFFAOYNA-N
SMILES: O.NC(CC1=CC=C(C=C1)[N+]([O-])=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)