missing translation for 'onlineSavingsMsg'
Learn More

4-(n-Undecyloxy)benzoic acid, 98%, Thermo Scientific™

Product Code. 11456416
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456416 1 g 1g
11466416 5 g 5g
11476416 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11456416 Supplier Thermo Scientific Alfa Aesar Supplier No. B24727.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15872-44-3
Molecular Formula C18H28O3
Molecular Weight (g/mol) 292.419
MDL Number MFCD00043772
InChI Key NEJZHJHZOUISSH-UHFFFAOYSA-N
Synonym 4-undecyloxy benzoic acid, 4-undecyloxybenzoic acid, 4-n-undecyloxybenzoic acid, p-undecyloxybenzoic acid, pubchem3929, 4-undecyloxybenzoicacid, acmc-209diw, 4-undecyloxy benzoic acid #, benzoic acid,4-undecyloxy, timtec-bb sbb008333
PubChem CID 519187
IUPAC Name 4-undecoxybenzoic acid
SMILES CCCCCCCCCCCOC1=CC=C(C=C1)C(=O)O

Specifications

Boiling Point 96°C to 137°C
Quantity 1 g
Formula Weight 292.42
Percent Purity 98%
Chemical Name or Material 4-(n-Undecyloxy)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.