accessibility menu, dialog, popup

UserName

CAS RN 15872-44-3

H3COOHO

CAS RN 15872-44-3

IUPAC Name: 4-(undecyloxy)benzoic acid
Synonyms: 4-(undecyloxy)benzoic acid
Molecular Weight (g/mol): 292.42
Molecular Formula: C18H28O3
InChi Key: NEJZHJHZOUISSH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCOC1=CC=C(C=C1)C(O)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results