missing translation for 'onlineSavingsMsg'
Learn More

4-n-Propylbenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals

Product Code. 11463917
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11463917 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11463917 Supplier Thermo Scientific Alfa Aesar Supplier No. A18354.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146949-07-7
Molecular Formula C9H11ClO2S
Molecular Weight (g/mol) 218.70
MDL Number MFCD00041508
InChI Key LEFGAGRZHLNPLS-UHFFFAOYSA-N
Synonym 4-n-propylbenzenesulfonyl chloride, 4-propylbenzene-1-sulfonyl chloride, 4-propyl-benzenesulfonylchloride, 4-propyl-benzenesulfonyl chloride, benzenesulfonyl chloride, 4-propyl, p-propylbenzenesulfonyl chloride, 4-n-propylbenzenesulfonylchloride, acmc-209cxt, 4-propylphenylsulfonyl chloride, 4-propylphenylsulphonyl chloride
PubChem CID 2737222
IUPAC Name 4-propylbenzenesulfonyl chloride
SMILES CCCC1=CC=C(C=C1)S(Cl)(=O)=O

Specifications

Melting Point 20°C to 22°C
Density 1.223
Boiling Point 275°C to 276°C
Refractive Index 1.5375
Quantity 25 g
UN Number UN3265
Beilstein 5511216
Sensitivity Moisture sensitive
Formula Weight 218.7
Percent Purity 95%
Chemical Name or Material 4-n-Propylbenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.