CAS RN 146949-07-7
IUPAC Name:
4-propylbenzene-1-sulfonyl chloride
Synonyms:
4-propylbenzenesulfonyl chloride
Molecular Weight (g/mol):
218.7
Molecular Formula:
C9H11ClO2S
InChi Key:
LEFGAGRZHLNPLS-UHFFFAOYSA-N
SMILES:
CCCC1=CC=C(C=C1)S(Cl)(=O)=O
1
–
1
of
1
results
1
4-n-Propylbenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals
CAS: 146949-07-7 Molecular Formula: C9H11ClO2S Molecular Weight (g/mol): 218.70 MDL Number: MFCD00041508 InChI Key: LEFGAGRZHLNPLS-UHFFFAOYSA-N Synonym: 4-n-propylbenzenesulfonyl chloride,4-propylbenzene-1-sulfonyl chloride,4-propyl-benzenesulfonylchloride,4-propyl-benzenesulfonyl chloride,benzenesulfonyl chloride, 4-propyl,p-propylbenzenesulfonyl chloride,4-n-propylbenzenesulfonylchloride,acmc-209cxt,4-propylphenylsulfonyl chloride,4-propylphenylsulphonyl chloride PubChem CID: 2737222 IUPAC Name: 4-propylbenzenesulfonyl chloride SMILES: CCCC1=CC=C(C=C1)S(Cl)(=O)=O