missing translation for 'onlineSavingsMsg'
Learn More

4-(N-Methylaminocarbonyl)phenylboronic acid, 95%, Thermo Scientific™

Product Code. 15206058 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15206058 250 mg 250mg
15296048 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15206058 Supplier Thermo Scientific Acros Supplier No. 454412500

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 121177-82-0
Molecular Formula C8H10BNO3
Molecular Weight (g/mol) 178.98
InChI Key UWKSYZHFTGONHY-UHFFFAOYSA-N
Synonym 4-n-methylaminocarbonyl phenylboronic acid, 4-methylcarbamoyl phenylboronic acid, 4-methylcarbamoyl phenyl boronic acid, 4-methylcarbamoyl benzeneboronic acid, boronic acid, 4-methylamino carbonyl phenyl, boronic acid,b-4-methylamino carbonyl phenyl, 4-methylamino carbonyl phenylboronic acid, 4-n-methylaminocarbonyl benzeneboronic acid, zlchem 836
PubChem CID 2773526
IUPAC Name [4-(methylcarbamoyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C(=O)NC)(O)O

Specifications

Melting Point 183.0°C to 186.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Quantity 250 mg
Formula Weight 178.98
Percent Purity 95%
Chemical Name or Material 4-(N-Methylaminocarbonyl)phenylboronic acid
compliance-icons
Product Identifier
  • 4-(N-Methylaminocarbonyl)phenylboronic acid, 95%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.