accessibility menu, dialog, popup

UserName

CH3HNOBOHHO

CAS RN 121177-82-0

IUPAC Name: [4-(methylcarbamoyl)phenyl]boronic acid
Synonyms: 4-(methylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 178.98
Molecular Formula: C8H10BNO3
InChi Key: UWKSYZHFTGONHY-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC=C(C=C1)B(O)O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)