missing translation for 'onlineSavingsMsg'
Learn More

4-Methylumbelliferyl a-L-Idopyranosiduronic Acid, Sodium, TRC

Product Code. 30007120
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
25 mg
Unit Size:
2.5mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007120 25 mg 25mg
30007004 5 mg 5mg
30004680 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007120 Supplier Toronto Research Chemicals Supplier No. TRCM33470125MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89157-94-8
Molecular Formula C16 H15 O9 . Na
Molecular Weight (g/mol) 374.27
Synonym 4-Methylumbelliferyl alpha-L-Iduronic Acid, Sodium
IUPAC Name sodium;(2R,3S,4S,5R,6S)-3,4,5-trihydroxy-6-(4-methyl-2-oxochromen-7-yl)oxyoxane-2-carboxylate
SMILES [Na+].CC1=CC(=O)Oc2cc(O[C@@H]3O[C@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)[O-])ccc12

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H16O9.Na/c1-6-4-10(17)24-9-5-7(2-3-8(6)9)23-16-13(20)11(18)12(19)14(25-16)15(21)22;/h2-5,11-14,16,18-20H,1H3,(H,21,22);/q;+1/p-1/t11-,12-,13+,14+,16+;/m0./s1
Formula Weight 374.0614
Percent Purity >95%
Chemical Name or Material 4-Methylumbelliferyl a-L-Idopyranosiduronic Acid, Sodium
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.