accessibility menu, dialog, popup

UserName

CH3OOOOOOHOHOHHONa

CAS RN 89157-94-8

IUPAC Name: 3,4,5-trihydroxy-6-[(4-methyl-2-oxo-2H-chromen-7-yl)oxy]oxane-2-carboxylic acid sodium
Synonyms: 3,4,5-trihydroxy-6-[(4-methyl-2-oxochromen-7-yl)oxy]oxane-2-carboxylic acid sodium
Molecular Weight (g/mol): 375.29
Molecular Formula: C16H16NaO9
InChi Key: IPAMIKZBFDASBC-UHFFFAOYNA-N
SMILES: [Na].CC1=CC(=O)OC2=CC(OC3OC(C(O)C(O)C3O)C(O)=O)=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results