missing translation for 'onlineSavingsMsg'
Learn More

4-(Methylthio)phenyl isocyanate, 97%, Thermo Scientific™

Product Code. 11318715
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11318715 1 g 1g
11328715 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11318715 Supplier Thermo Scientific Alfa Aesar Supplier No. L12263.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1632-84-4
Molecular Formula C8H7NOS
Molecular Weight (g/mol) 165.21
MDL Number MFCD00013878
InChI Key QNKQBDZVEKZFBN-UHFFFAOYSA-N
Synonym 4-methylthio phenyl isocyanate, 1-isocyanato-4-methylthio benzene, 1-isocyanato-4-methylsulfanyl benzene, 4-methylsulfanyl phenyl isocyanate, 4-methylthio phenylisocyanate, 4-methylthiophenyl isocyanate, 4-methylthiobenzenisocyanate, acmc-20amu4, p-methylthiophenylisocyanate, 4-methylthiophenylisocyanate
PubChem CID 2733316
IUPAC Name 1-isocyanato-4-methylsulfanylbenzene
SMILES CSC1=CC=C(C=C1)N=C=O

Specifications

Boiling Point 104°C to 105°C (0.5 mmHg)
Flash Point 104°C (219°F)
Odor Pungent
Quantity 1 g
UN Number UN2206
Beilstein 1936612
Formula Weight 165.22
Percent Purity 97%
Chemical Name or Material 4-(Methylthio)phenyl isocyanate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.