accessibility menu, dialog, popup

UserName

H3CSNCO

CAS RN 1632-84-4

IUPAC Name: 1-isocyanato-4-(methylsulfanyl)benzene
Synonyms: 1-isocyanato-4-(methylsulfanyl)benzene
Molecular Weight (g/mol): 165.21
Molecular Formula: C8H7NOS
InChi Key: QNKQBDZVEKZFBN-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C=C1)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)