missing translation for 'onlineSavingsMsg'
Learn More

(4-methyl-3,4-dihydro-2h-1,4-benzoxazin-2-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10265454
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10265454 1 g Amber Glass Bottle 1g
10726905 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10265454 Supplier Thermo Scientific Maybridge Supplier No. CC29813DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 282520-55-2
Molecular Formula C10H14N2O
Molecular Weight (g/mol) 178.24
MDL Number MFCD11841068
InChI Key VPYSMSLDVAQICD-UHFFFAOYNA-N
Synonym 4-methyl-3,4-dihydro-2h-1,4-benzoxazin-2-yl methylamine, 2-aminomethyl-3,4-dihydro-4-methyl-2h-1,4-benzoxazine, 4-methyl-2,3-dihydro-1,4-benzoxazin-2-yl methanamine, 4-methyl-3,4-dihydro-2h-benzo b 1,4 oxazin-2-yl methanamine, 1-4-methyl-2,3-dihydro-1,4-benzoxazin-2-yl methanamine, 4-methyl-2h,3h-benzo e 1,4-oxazin-2-yl methylamine, 3,4-dihydro-4-methyl-2h-1,4-benzoxazin-2-yl methylamine, 1-4-methyl-3,4-dihydro-2h-1,4-benzoxazin-2-yl methanamine, c-4-methyl-3,4-dihydro-2h-benzo 1,4 oxazin-2-yl-methylamine
PubChem CID 15550374
IUPAC Name (4-methyl-2,3-dihydro-1,4-benzoxazin-2-yl)methanamine
SMILES CN1CC(CN)OC2=CC=CC=C12

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 178.23
Percent Purity 97%
Chemical Name or Material (4-Methyl-3, 4-dihydro-2H-1, 4-benzoxazin-2-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.