accessibility menu, dialog, popup

UserName

CH3NOH2N

CAS RN 282520-55-2

IUPAC Name: 1-(4-methyl-3,4-dihydro-2H-1,4-benzoxazin-2-yl)methanamine
Synonyms: 1-(4-methyl-2,3-dihydro-1,4-benzoxazin-2-yl)methanamine
Molecular Weight (g/mol): 178.24
Molecular Formula: C10H14N2O
InChi Key: VPYSMSLDVAQICD-UHFFFAOYNA-N
SMILES: CN1CC(CN)OC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results