missing translation for 'onlineSavingsMsg'
Learn More

4-Methyl-1-naphthoyl chloride, 98%, Thermo Scientific™

Product Code. 15452339
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
Product Code. Quantity unitSize
15452339 25 g 25g
15432339 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15452339

Brand: Thermo Scientific Alfa Aesar H64917.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87700-67-2
Molecular Formula C12H9ClO
Molecular Weight (g/mol) 204.653
MDL Number MFCD12025362
InChI Key IXGMHYHWSPDNNS-UHFFFAOYSA-N
Synonym 1-naphthalenecarbonylchloride, 4-methyl, 1-naphthalenecarbonyl chloride, 4-methyl, 4-methyl-1-naphthalenecarbonyl chloride, 1-naphthalenecarbonyl chloride, 4-methyl-9ci, 4-methyl-1-napthoyl chloride, 4-methyl-1-naphthoyl chloride, 4-methyl-naphthalene-1-carbonyl chloride
PubChem CID 24887412
IUPAC Name 4-methylnaphthalene-1-carbonyl chloride
SMILES CC1=CC=C(C2=CC=CC=C12)C(=O)Cl

Specifications

Color Yellow
Quantity 25 g
UN Number UN3261
Beilstein 3247818
Formula Weight 204.65
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Methyl-1-naphthoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.