accessibility menu, dialog, popup

UserName

CAS RN 87700-67-2

CH3OCl

CAS RN 87700-67-2

IUPAC Name: 4-methylnaphthalene-1-carbonyl chloride
Synonyms: 4-methylnaphthalene-1-carbonyl chloride
Molecular Weight (g/mol): 204.65
Molecular Formula: C12H9ClO
InChi Key: IXGMHYHWSPDNNS-UHFFFAOYSA-N
SMILES: CC1=C2C=CC=CC2=C(C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results