missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxyphenylmagnesium bromide, 1.0 M in 2-MeTHF

Product Code. 15471036
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15471036 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15471036 Supplier Thermo Scientific Alfa Aesar Supplier No. H54317.AE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 13139-86-1 | C7H7BrMgO | 211.34 g/mol

4-Methoxyphenylmagnesium bromide is applied in Grignard-reactions as a reagent for the introduction of the 4-methoxyphenyl group.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Methoxyphenylmagnesium bromide is applied in Grignard-reactions as a reagent for the introduction of the 4-methoxyphenyl group.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. I
TRUSTED_SUSTAINABILITY

Specifications

CAS 13139-86-1
Molecular Formula C7H7BrMgO
MDL Number MFCD00013515
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym 4-methoxyphenylmagnesium bromide, 4-methoxyphenylmagnesium bromide solution, bromo 4-methoxyphenyl magnesium, p-methoxyphenylmagnesium bromide, 4-methoxyphenyl magnesium bromide, 4-methoxyphenymagnesium bromide, 4-methoxy phenylmagnesiumbromide, p-methoxyphenyl magnesium bromide, p-methoxyphenyl-magnesium bromide, 4-methoxy-phenyl-magnesiumbromide, grignard reagent
Solubility Information It reacts with water.
InChI Key JKXAZEXIOPFJSC-UHFFFAOYSA-M
SMILES COC1=CC=C([Mg]Br)C=C1
IUPAC Name magnesium;methoxybenzene;bromide
Molecular Weight (g/mol) 211.34
PubChem CID 2734898
Formula Weight 211.34
Concentration or Composition (by Analyte or Components) 1.0M in 2-MeTHF
Chemical Name or Material 4-Methoxyphenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.