missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxycinnamic Acid, TRC

Product Code. 30480931
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480931 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30480931 Supplier Toronto Research Chemicals Supplier No. TRCM2621501G
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 830-09-1
Molecular Formula C10 H10 O3
Quantity 1 g
Synonym 2-Propenoic acid, 3-(4-methoxyphenyl)-, Cinnamic acid, p-methoxy- (6CI,8CI), 3-(4-Methoxyphenyl)-2-propenoic acid, 3-(4-Methoxyphenyl)acrylic acid, 3-(4-Methoxyphenyl)propenoic acid, 4-Methoxycinnamic acid, 4-Methyoxycinnamic acid, 4'-Methoxycinnamic acid, Bernel hydro, NSC 5303, NSC 623437, O-Methyl-p-coumaric acid, p-Methoxycinnamic acid, (EZ)-4-Methoxycinnamic acid
InChI Formula InChI=1S/C10H10O3/c1-13-9-5-2-8(3-6-9)4-7-10(11)12/h2-7H,1H3,(H,11,12)/b7-4+
SMILES COc1ccc(\C=C\C(=O)O)cc1
IUPAC Name (E)-3-(4-methoxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol) 178.18
Formula Weight 178.063
Chemical Name or Material 4-Methoxycinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.