missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxycinnamic acid, 99%

Product Code. 15495506
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495506 100 g 100g
15446618 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15495506 Supplier Thermo Scientific Alfa Aesar Supplier No. H60144.22
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 830-09-1 | C10H10O3 | 178.187 g/mol

It exerts anti-hyperglycemic/ hypoglycemic effect by stimulating insulin secretion from pancreas. It could be developed into a new potential therapeutic agent used in type 2 diabetic patients.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It exerts anti-hyperglycemic/ hypoglycemic effect by stimulating insulin secretion from pancreas. It could be developed into a new potential therapeutic agent used in type 2 diabetic patients.

Solubility
Soluble in dimethyl sulfoxide and methanol.

Notes
Store in cool dry conditions. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 830-09-1
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00004398
InChI Key AFDXODALSZRGIH-QPJJXVBHSA-N
Synonym 4-methoxycinnamic acid, p-methoxycinnamic acid, 3-4-methoxyphenyl acrylic acid, trans-4-methoxycinnamic acid, 4-methoxycinnamate, para-methoxycinnamic acid, o-methyl-p-coumaric acid, cinnamic acid, p-methoxy, e-3-4-methoxyphenyl acrylic acid, e-3-4-methoxyphenyl-2-propenoic acid
PubChem CID 699414
IUPAC Name (E)-3-(4-methoxyphenyl)prop-2-enoic acid
SMILES COC1=CC=C(C=C1)C=CC(=O)O

Specifications

Melting Point 170°C to 173°C
Quantity 100 g
Solubility Information Soluble in dimethyl sulfoxide and methanol.
Formula Weight 178.18
Percent Purity 99%
Chemical Name or Material 4-Methoxycinnamic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.