missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethyl)benzoyl chloride, 97%

Product Code. 11486266
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486266 5 g 5g
11476266 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486266 Supplier Thermo Scientific Alfa Aesar Supplier No. B23072.06
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 98187-18-9 | C9H6ClF3O2 | 238.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98187-18-9
Molecular Formula C9H6ClF3O2
Molecular Weight (g/mol) 238.59
MDL Number MFCD01631576
InChI Key PBKYPTQBAQQVPT-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethyl benzoyl chloride, 3-trifluoromethyl-p-anisoyl chloride, 5-chlorocarbonyl-2-methoxybenzotrifluoride, 4methoxy-3-trifluoromethylbenzoyl chloride, 4-methoxy-5-trifluoromethylbenzoyl chloride, 3-trifluoromethyl-4-methoxy-benzoyl chloride, 4-methoxy-3-trifluoromethyl benzoylchloride, 4-methoxy-3-trifluromethyl benzoyl chloride, 4-methoxy-3-trifluoromethyl-benzoyl chloride
PubChem CID 2775298
IUPAC Name 4-methoxy-3-(trifluoromethyl)benzoyl chloride
SMILES COC1=CC=C(C=C1C(F)(F)F)C(Cl)=O

Specifications

Refractive Index 1.515
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 238.59
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-(trifluoromethyl)benzoyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.