accessibility menu, dialog, popup

UserName

CH3OFFFClO

CAS RN 98187-18-9

IUPAC Name: 4-methoxy-3-(trifluoromethyl)benzoyl chloride
Synonyms: 4-methoxy-3-(trifluoromethyl)benzoyl chloride
Molecular Weight (g/mol): 238.59
Molecular Formula: C9H6ClF3O2
InChi Key: PBKYPTQBAQQVPT-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1C(F)(F)F)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results