missing translation for 'onlineSavingsMsg'
Learn More

4-Methoxy-3-(trifluoromethoxy)benzoyl chloride, 97%, Thermo Scientific™

Product Code. p-7032190
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15418535 250 mg 250mg
15408535 1 g 1g
2 options
This item is not returnable. View return policy

Product Code. 15418535

Brand: Thermo Scientific Alfa Aesar H33563.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1070774-23-0
Molecular Formula C9H6ClF3O3
Molecular Weight (g/mol) 254.59
MDL Number MFCD18398465
InChI Key HWYUFLOSGDZCMG-UHFFFAOYSA-N
Synonym 4-methoxy-3-trifluoromethoxy benzoyl chloride, 4-methoxy-3-trifluoromethoxy-benzoyl chloride, benzoyl chloride, 4-methoxy-3-trifluoromethoxy
PubChem CID 66523505
IUPAC Name 4-methoxy-3-(trifluoromethoxy)benzoyl chloride
SMILES COC1=C(OC(F)(F)F)C=C(C=C1)C(Cl)=O

Specifications

Quantity 250 mg
UN Number UN3265
Sensitivity Air sensitive
Formula Weight 254.59
Percent Purity 97%
Chemical Name or Material 4-Methoxy-3-(trifluoromethoxy)benzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.