CAS RN 1070774-23-0
IUPAC Name:
4-methoxy-3-(trifluoromethoxy)benzoyl chloride
Synonyms:
4-methoxy-3-(trifluoromethoxy)benzoyl chloride
Molecular Weight (g/mol):
254.59
Molecular Formula:
C9H6ClF3O3
InChi Key:
HWYUFLOSGDZCMG-UHFFFAOYSA-N
SMILES:
COC1=C(OC(F)(F)F)C=C(C=C1)C(Cl)=O
1
–
1
of
1
results
1
4-Methoxy-3-(trifluoromethoxy)benzoyl chloride, 97%, Thermo Scientific™
CAS: 1070774-23-0 Molecular Formula: C9H6ClF3O3 Molecular Weight (g/mol): 254.59 MDL Number: MFCD18398465 InChI Key: HWYUFLOSGDZCMG-UHFFFAOYSA-N Synonym: 4-methoxy-3-trifluoromethoxy benzoyl chloride,4-methoxy-3-trifluoromethoxy-benzoyl chloride,benzoyl chloride, 4-methoxy-3-trifluoromethoxy PubChem CID: 66523505 IUPAC Name: 4-methoxy-3-(trifluoromethoxy)benzoyl chloride SMILES: COC1=C(OC(F)(F)F)C=C(C=C1)C(Cl)=O