missing translation for 'onlineSavingsMsg'
Learn More

4-Keto Retinol (~90%), TRC

Product Code. 30394726
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394726 25 mg -
30394924 5 mg -
30396654 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30394726 Supplier Toronto Research Chemicals Supplier No. TRCK20500025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62702-55-0
Molecular Formula C20 H28 O2
Molecular Weight (g/mol) 300.44
Synonym Retin-4-one, 15-hydroxy-, 2-Cyclohexen-1-one, 3-(9-hydroxy-3,7-dimethyl-1,3,5,7-nonatetraenyl)-2,4,4-trimethyl- (6CI), Retinol, 4-oxo- (9CI), 15-Hydroxyretin-4-one, 4-Oxoretinol, all-trans-4-Oxoretinol, 4-Ketoretinol
IUPAC Name 3-[(1E,3E,5E,7E)-9-hydroxy-3,7-dimethylnona-1,3,5,7-tetraenyl]-2,4,4-trimethylcyclohex-2-en-1-one
SMILES C\C(=C/CO)\C=C\C=C(/C)\C=C\C1=C(C)C(=O)CCC1(C)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H28O2/c1-15(7-6-8-16(2)12-14-21)9-10-18-17(3)19(22)11-13-20(18,4)5/h6-10,12,21H,11,13-14H2,1-5H3/b8-6+,10-9+,15-7+,16-12+
Formula Weight 300.2089
Chemical Name or Material 4-Ketoretinol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.