CAS RN 62702-55-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Keto Retinol (~90%), TRC
CAS: 62702-55-0 Molecular Formula: C20 H28 O2 Molecular Weight (g/mol): 300.44 Synonym: Retin-4-one, 15-hydroxy-,2-Cyclohexen-1-one, 3-(9-hydroxy-3,7-dimethyl-1,3,5,7-nonatetraenyl)-2,4,4-trimethyl- (6CI),Retinol, 4-oxo- (9CI),15-Hydroxyretin-4-one,4-Oxoretinol,all-trans-4-Oxoretinol,4-Ketoretinol IUPAC Name: 3-[(1E,3E,5E,7E)-9-hydroxy-3,7-dimethylnona-1,3,5,7-tetraenyl]-2,4,4-trimethylcyclohex-2-en-1-one SMILES: C\C(=C/CO)\C=C\C=C(/C)\C=C\C1=C(C)C(=O)CCC1(C)C