missing translation for 'onlineSavingsMsg'
Learn More

4-Keto 13-cis-Retinoic Acid Methyl Ester, TRC

Product Code. 30397695
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397695 2.5 mg 2.5mg
30397510 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397695 Supplier Toronto Research Chemicals Supplier No. TRCK2049752.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71748-57-7
Molecular Formula C21 H28 O3
Molecular Weight (g/mol) 328.45
Synonym 13-cis-4-Oxo-retinoic acid methyl ester
IUPAC Name methyl (2Z,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethyl-3-oxocyclohexen-1-yl)nona-2,4,6,8-tetraenoate
SMILES COC(=O)\C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)C(=O)CCC1(C)C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C21H28O3/c1-15(8-7-9-16(2)14-20(23)24-6)10-11-18-17(3)19(22)12-13-21(18,4)5/h7-11,14H,12-13H2,1-6H3/b9-7+,11-10+,15-8+,16-14-
Formula Weight 328.2038
Chemical Name or Material 4-Keto 13-cis-Retinoic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.