CAS RN 71748-57-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Keto 13-cis-Retinoic Acid Methyl Ester, TRC
CAS: 71748-57-7 Molecular Formula: C21 H28 O3 Molecular Weight (g/mol): 328.45 Synonym: 13-cis-4-Oxo-retinoic acid methyl ester IUPAC Name: methyl (2Z,4E,6E,8E)-3,7-dimethyl-9-(2,6,6-trimethyl-3-oxocyclohexen-1-yl)nona-2,4,6,8-tetraenoate SMILES: COC(=O)\C=C(\C)/C=C/C=C(\C)/C=C/C1=C(C)C(=O)CCC1(C)C