missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ 4-IPP
GREENER_CHOICE

Product Code. 18074502
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18074502

Brand: Tocris Bioscience™ 3429/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Inhibitor of macrophage migration inhibitory factor (MIF); suicide substrate

Inhibitor of macrophage migration inhibitory factor (MIF). Acts as a suicide substrate to inactivate MIF catalytic and biological functions; inhibits MIF-associated liver enzyme activity in vivo. Also inhibits migration and anchorage independence of human lung adenocarcinoma cell lines in vitro.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41270-96-6
Molecular Formula C10H7IN2

Specifications

Quantity 50 mg
Formula Weight 282.08
Purity >99%
Molecular Weight (g/mol) 282.084
Chemical Name or Material 4-IPP
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.