CAS RN 41270-96-6
CAS RN 41270-96-6
IUPAC Name:
4-iodo-6-phenylpyrimidine
Synonyms:
4-iodo-6-phenylpyrimidine
Molecular Weight (g/mol):
282.08
Molecular Formula:
C10H7IN2
InChi Key:
ZTCJXHNJVLUUMR-UHFFFAOYSA-N
SMILES:
IC1=NC=NC(=C1)C1=CC=CC=C1