missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzyl alcohol, 97%

Product Code. 11324166
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11324166 1 g 1g
11334166 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11324166 Supplier Thermo Scientific Alfa Aesar Supplier No. L19278.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18282-51-4 | C7H7IO | 234.036 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18282-51-4
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD01732720
InChI Key CNQRHSZYVFYOIE-UHFFFAOYSA-N
Synonym 4-iodobenzyl alcohol, 4-iodophenyl methanol, p-iodobenzyl alcohol, benzenemethanol, 4-iodo, benzyl alcohol, p-iodo, 4-iodobenzylalcohol, p-iodobenzylalcohol, 4-iodobenzyl-alcohol, 4-iodobenzyl zlcohol, p-lodo-benzyl alcohol
PubChem CID 29012
IUPAC Name (4-iodophenyl)methanol
SMILES C1=CC(=CC=C1CO)I

Specifications

Melting Point 72°C to 74°C
Quantity 1 g
Beilstein 1931621
Sensitivity Light sensitive
Formula Weight 234.04
Percent Purity 97%
Chemical Name or Material 4-Iodobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.