CAS RN 18282-51-4
IUPAC Name:
(4-iodophenyl)methanol
Synonyms:
benzenemethanol, 4-iodo-
Molecular Weight (g/mol):
234.04
Molecular Formula:
C7H7IO
InChi Key:
CNQRHSZYVFYOIE-UHFFFAOYSA-N
SMILES:
OCC1=CC=C(I)C=C1
1
–
2
of
2
results
1
4-Iodobenzyl alcohol, 97%
CAS: 18282-51-4 Molecular Formula: C7H7IO Molecular Weight (g/mol): 234.036 MDL Number: MFCD01732720 InChI Key: CNQRHSZYVFYOIE-UHFFFAOYSA-N Synonym: 4-iodobenzyl alcohol,4-iodophenyl methanol,p-iodobenzyl alcohol,benzenemethanol, 4-iodo,benzyl alcohol, p-iodo,4-iodobenzylalcohol,p-iodobenzylalcohol,4-iodobenzyl-alcohol,4-iodobenzyl zlcohol,p-lodo-benzyl alcohol PubChem CID: 29012 IUPAC Name: (4-iodophenyl)methanol SMILES: C1=CC(=CC=C1CO)I