missing translation for 'onlineSavingsMsg'
Learn More

4-(Iodoacetylamino)phenol, TRC

Product Code. 30397649
Change view
Click to view available options
Quantity:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397649 50 mg
30406485 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397649 Supplier Toronto Research Chemicals Supplier No. TRCI53525050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53527-05-2
Molecular Formula C8H8INO2
Molecular Weight (g/mol) 277.06
Synonym 4’-Hydroxy-2-iodoacetanilide, N-(4-Hydroxyphenyl)-2-iodo-acetamide
IUPAC Name N-(4-hydroxyphenyl)-2-iodoacetamide
SMILES Oc1ccc(NC(=O)CI)cc1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C8H8INO2/c9-5-8(12)10-6-1-3-7(11)4-2-6/h1-4,11H,5H2,(H,10,12)
Formula Weight 276.96
Chemical Name or Material 4-(Iodoacetylamino)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.