CAS RN 53527-05-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(Iodoacetylamino)phenol, TRC
CAS: 53527-05-2 Molecular Formula: C8H8INO2 Molecular Weight (g/mol): 277.06 Synonym: 4’-Hydroxy-2-iodoacetanilide,N-(4-Hydroxyphenyl)-2-iodo-acetamide IUPAC Name: N-(4-hydroxyphenyl)-2-iodoacetamide SMILES: Oc1ccc(NC(=O)CI)cc1