missing translation for 'onlineSavingsMsg'
Learn More

4’-Hydroxy rac-Kavain, TRC

Product Code. 30405744
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30405744 5 mg
30390659 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30405744 Supplier Toronto Research Chemicals Supplier No. TRCH9445205MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1401441-87-9
Molecular Formula C14H14O4
Molecular Weight (g/mol) 246.26
Synonym 5,6-Dihydro-6-[(1E)-2-(4-hydroxyphenyl)ethenyl]-4-methoxy-2H-pyran-2-one
IUPAC Name 2-[(E)-2-(4-hydroxyphenyl)ethenyl]-4-methoxy-2,3-dihydropyran-6-one
SMILES O=C1C=C(OC)CC(/C=C/C2=CC=C(O)C=C2)O1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H14O4/c1-17-13-8-12(18-14(16)9-13)7-4-10-2-5-11(15)6-3-10/h2-7,9,12,15H,8H2,1H3/b7-4+
Formula Weight 246.0892
Chemical Name or Material 4'-Hydroxy rac-Kavain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.