CAS RN 1401441-87-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4’-Hydroxy rac-Kavain, TRC
CAS: 1401441-87-9 Molecular Formula: C14H14O4 Molecular Weight (g/mol): 246.26 Synonym: 5,6-Dihydro-6-[(1E)-2-(4-hydroxyphenyl)ethenyl]-4-methoxy-2H-pyran-2-one IUPAC Name: 2-[(E)-2-(4-hydroxyphenyl)ethenyl]-4-methoxy-2,3-dihydropyran-6-one SMILES: O=C1C=C(OC)CC(/C=C/C2=CC=C(O)C=C2)O1