missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxy Pyrimethanil-d4, TRC

Product Code. 30406453
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406453 1 mg
30404487 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30406453 Supplier Toronto Research Chemicals Supplier No. TRCH9532071MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1794897-91-8
Molecular Formula C12H9D4N3O
Molecular Weight (g/mol) 219.28
Synonym 2-(4-Hydroxyanilino)-4,6-dimethylpyrimidine-d4, 4-[(4,6-Dimethyl-2-pyrimidinyl)amino]phenol-d4
SMILES OC(C([2H])=C1[2H])=C([2H])C([2H])=C1NC2=NC(C)=CC(C)=N2

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C12H13N3O/c1-8-7-9(2)14-12(13-8)15-10-3-5-11(16)6-4-10/h3-7,16H,1-2H3,(H,13,14,15)/i3D,4D,5D,6D
Formula Weight 219.131
Chemical Name or Material 4'-Hydroxy Pyrimethanil-d4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.