CAS RN 1794897-91-8
CAS RN 1794897-91-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4'-Hydroxy Pyrimethanil-d4, TRC
CAS: 1794897-91-8 Molecular Formula: C12H9D4N3O Molecular Weight (g/mol): 219.28 Synonym: 2-(4-Hydroxyanilino)-4,6-dimethylpyrimidine-d4,4-[(4,6-Dimethyl-2-pyrimidinyl)amino]phenol-d4 SMILES: OC(C([2H])=C1[2H])=C([2H])C([2H])=C1NC2=NC(C)=CC(C)=N2