missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-Alpha1-[[[6-(1-methyl-3-phenylpropoxy)hexyl]amino]methyl]-1,3-benzenedimethanol (Salmeterol Impurity), TRC

Product Code. 30134850
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134850 25 mg 25mg
17991495 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30134850 Supplier Toronto Research Chemicals Supplier No. TRCH94817025MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108928-81-0
Molecular Formula C25 H37 N O4
Molecular Weight (g/mol) 415.57
Synonym 1,3-Benzenedimethanol, 4-hydroxy-α1-[[[6-(1-methyl-3-phenylpropoxy)hexyl]amino]methyl]-, 4-{1-Hydroxy-2-[6-(4-phenylbutan-2-yloxy)hexylamino]ethyl}-2-(hydroxymethyl)phenol, 1-[4-Hydroxy-3-(hydroxymethyl)-phenyl]-2-[[6-(1-methyl-3-phenylpropoxy)hexyl]-amino]ethanol, Salmeterol Related Compound B (USP), Salmeterol Xinafoate Imp. E (EP), GR 58583X, USP Salmeterol Related Compound B, Ph Eur Salmeterol Impurity E, GR 58583X
IUPAC Name 2-(hydroxymethyl)-4-[1-hydroxy-2-[6-(4-phenylbutan-2-yloxy)hexylamino]ethyl]phenol
SMILES CC(CCc1ccccc1)OCCCCCCNCC(O)c2ccc(O)c(CO)c2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C25H37NO4/c1-20(11-12-21-9-5-4-6-10-21)30-16-8-3-2-7-15-26-18-25(29)22-13-14-24(28)23(17-22)19-27/h4-6,9-10,13-14,17,20,25-29H,2-3,7-8,11-12,15-16,18-19H2,1H3
Formula Weight 415.2723
Chemical Name or Material 4-{1-Hydroxy-2-[6-(4-phenylbutan-2-yloxy)hexylamino]ethyl}-2-(hydroxymethyl)phenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.