missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol, 97+%, Thermo Scientific™

Product Code. 11330548
Change view
Click to view available options
Quantity:
0.5 g
2 g
10 g
Unit Size:
0.5g
10g
2g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11330548 2 g 2g
11340548 0.5 g 0.5g
11350548 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11330548 Supplier Thermo Scientific Alfa Aesar Supplier No. 043873.04
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 368-54-7
Molecular Formula C5H3F3N2OS
Molecular Weight (g/mol) 196.147
MDL Number MFCD00112978
InChI Key YZAOETRYQWFEOY-UHFFFAOYSA-N
Synonym 4-hydroxy-6-trifluoromethyl pyrimidine-2-thiol, 2-mercapto-6-trifluoromethyl pyrimidin-4-ol, 2-sulfanyl-6-trifluoromethyl pyrimidin-4-ol, 2-sulfanyl-6-trifluoromethyl-4-pyrimidinol, 4-hydroxy-2-mercapto-6-trifluoromethyl pyrimidine, 6-trifluoromethyl-2-thiouracil, 2-thioxo-6-trifluoromethyl-2,3-dihydropyrimidin-4 1h-one, 4-hydroxy-2-mercapto-6-trifluoromethylpyrimidine, uracil, 2-thio-6-trifluoromethyl, 2-mercapto-6-trifluoromethyl-pyrimidin-4-ol
PubChem CID 1239796
IUPAC Name 2-sulfanylidene-6-(trifluoromethyl)-1H-pyrimidin-4-one
SMILES C1=C(NC(=S)NC1=O)C(F)(F)F

Specifications

Melting Point 251°C
Quantity 2 g
Formula Weight 196.1
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 4-Hydroxy-6- (trifluoromethyl)pyrimidine-2-thiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.