accessibility menu, dialog, popup

UserName

FFFONHSHN

CAS RN 368-54-7

IUPAC Name: 2-sulfanylidene-6-(trifluoromethyl)-1,2,3,4-tetrahydropyrimidin-4-one
Synonyms: 2-sulfanylidene-6-(trifluoromethyl)-1,3-dihydropyrimidin-4-one
Molecular Weight (g/mol): 196.15
Molecular Formula: C5H3F3N2OS
InChi Key: YZAOETRYQWFEOY-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC(=O)NC(=S)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results