missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-nitrophenylarsonic Acid, TRC

Product Code. 30398799
Change view
Click to view available options
Quantity:
2.5 g
25 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398799 25 g -
30391878 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398799 Supplier Toronto Research Chemicals Supplier No. TRCH94868025G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-19-7
Molecular Formula C6 H6 As N O6
Molecular Weight (g/mol) 263.04
Synonym Arsonic acid, As-(4-hydroxy-3-nitrophenyl)- (ACI), Arsonic acid, (4-hydroxy-3-nitrophenyl)- (9CI), As-(4-Hydroxy-3-nitrophenyl)arsonic acid (ACI), Benzenearsonic acid, 4-hydroxy-3-nitro- (6CI, 7CI, 8CI), 2-Nitro-1-hydroxybenzene-4-arsonic acid, 3-Nitro, 3-Nitro-10, 3-Nitro-20, 3-Nitro-4-hydroxybenzenearsonic acid, 3-Nitro-4-hydroxyphenylarsonic acid, 3-Nitro-50, 3-Nitro-80, 4-Hydroxy-3-nitrobenzenearsonic acid, 4-Hydroxy-3-nitrophenylarsonic acid, Aklomix-3, MeSH ID: D012406, NSC 2101, p-Hydroxy-m-nitrophenylarsonic acid, Ren-O-sal, Ristat, Roxarsone
IUPAC Name (4-hydroxy-3-nitrophenyl)arsonic acid
SMILES Oc1ccc(cc1[N+](=O)[O-])[As](=O)(O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C6H6AsNO6/c9-6-2-1-4(7(10,11)12)3-5(6)8(13)14/h1-3,9H,(H2,10,11,12)
Formula Weight 262.9411
Chemical Name or Material Roxarsone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.