accessibility menu, dialog, popup

UserName

OHNOOAsOHHOO

CAS RN 121-19-7

IUPAC Name: (4-hydroxy-3-nitrophenyl)arsonic acid
Synonyms: 3-nitro roxarsone ren-O-sal roxarson 罗沙胂 roxarsonum
Molecular Weight (g/mol): 263.04
Molecular Formula: C6H6AsNO6
InChi Key: XMVJITFPVVRMHC-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1[N+]([O-])=O)[As](O)(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results