missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Product Code. 15427555
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427555 250 mg 250mg
11362119 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15427555 Supplier Thermo Scientific Alfa Aesar Supplier No. H26127.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
4-Hydroxy-2-(trifluoromethyl)benzoic acid is used in pharm industry.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 320-32-1
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD01091007
InChI Key CSQAVQLFCBRQJM-UHFFFAOYSA-N
Synonym 4-hydroxy-2-trifluoromethyl benzoic acid, benzoic acid, 4-hydroxy-2-trifluoromethyl, pubchem13890, acmc-1ahif, 2-carboxy-5-hydroxybenzotrifluoride, 4-carboxy-3-trifluoromethyl phenol, 2-trifluoromethyl-4-hydroxybenzoic acid, 4-hydroxy-2-trifluoromethyl-benzoic acid, 4-hydroxy-2-trifluoromethyl benzoicacid
PubChem CID 2775111
IUPAC Name 4-hydroxy-2-(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C=C1O)C(F)(F)F)C(=O)O

Specifications

Melting Point 159°C to 161°C
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 206.12
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-2-(trifluoromethyl)benzoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.