accessibility menu, dialog, popup

UserName

OHFFFOHO

CAS RN 320-32-1

IUPAC Name: 4-hydroxy-2-(trifluoromethyl)benzoic acid
Synonyms: 4-hydroxy-2-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 206.12
Molecular Formula: C8H5F3O3
InChi Key: CSQAVQLFCBRQJM-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(C=C(O)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results