missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-phenylquinoline, TRC

Product Code. 30390068
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390068 50 mg -
30395230 500 mg 500mg
30405371 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390068 Supplier Toronto Research Chemicals Supplier No. TRCH11320550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1144-20-3
Molecular Formula C15H11NO
Molecular Weight (g/mol) 221.26
IUPAC Name 2-phenyl-1H-quinolin-4-one
SMILES O=C1C=C(Nc2ccccc12)c3ccccc3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H11NO/c17-15-10-14(11-6-2-1-3-7-11)16-13-9-5-4-8-12(13)15/h1-10H,(H,16,17)
Formula Weight 221.0841
Chemical Name or Material 4-Hydroxy-2-phenylquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.