CAS RN 1144-20-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Hydroxy-2-phenylquinoline, TRC
CAS: 1144-20-3 Molecular Formula: C15H11NO Molecular Weight (g/mol): 221.26 IUPAC Name: 2-phenyl-1H-quinolin-4-one SMILES: O=C1C=C(Nc2ccccc12)c3ccccc3