missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophthalonitrile, 98%

Product Code. p-7026567
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 11384719

Brand: Thermo Scientific Alfa Aesar B22624.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 65610-14-2 | C8H3FN2 | 146.124 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65610-14-2
Molecular Formula C8H3FN2
Molecular Weight (g/mol) 146.124
MDL Number MFCD00210430
InChI Key QQEKYCCJLSRLEC-UHFFFAOYSA-N
Synonym 4-fluorophthalonitrile, 1,2-benzenedicarbonitrile, 4-fluoro, 4-fluorophthalodinitrile, 1,2-dicyano-4-fluorobenzene, 4-fluoro-phthalonitrile, 2-cyano-5-fluorobenzonitrile, 4-fluorophthalonitrle, pubchem4027, acmc-1bb7s
PubChem CID 103423
IUPAC Name 4-fluorobenzene-1,2-dicarbonitrile
SMILES C1=CC(=C(C=C1F)C#N)C#N

Specifications

Melting Point 100°C to 102°C
Quantity 250 mg
UN Number UN3439
Beilstein 2443236
Formula Weight 146.12
Percent Purity 98%
Chemical Name or Material 4-Fluorophthalonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.