CAS RN 65610-14-2
CAS RN 65610-14-2
IUPAC Name:
4-fluorobenzene-1,2-dicarbonitrile
Synonyms:
4-fluorobenzene-1,2-dicarbonitrile
Molecular Weight (g/mol):
146.12
Molecular Formula:
C8H3FN2
InChi Key:
QQEKYCCJLSRLEC-UHFFFAOYSA-N
SMILES:
FC1=CC=C(C#N)C(=C1)C#N
1
–
1
of
1
results
1
4-Fluorophthalonitrile, 98%
CAS: 65610-14-2 Molecular Formula: C8H3FN2 Molecular Weight (g/mol): 146.124 MDL Number: MFCD00210430 InChI Key: QQEKYCCJLSRLEC-UHFFFAOYSA-N Synonym: 4-fluorophthalonitrile,1,2-benzenedicarbonitrile, 4-fluoro,4-fluorophthalodinitrile,1,2-dicyano-4-fluorobenzene,4-fluoro-phthalonitrile,2-cyano-5-fluorobenzonitrile,4-fluorophthalonitrle,pubchem4027,acmc-1bb7s PubChem CID: 103423 IUPAC Name: 4-fluorobenzene-1,2-dicarbonitrile SMILES: C1=CC(=C(C=C1F)C#N)C#N